Skip to Content
MilliporeSigma
All Photos(1)

Documents

272051

Sigma-Aldrich

1,3-Di-tert-butylbenzene

97%

Synonym(s):

1,3-Bis(1,1-dimethylethyl)benzene, 1,3-Ditert-butylbenzene, 4,6-Di(tert-butyl)benzene, m-Di-tert-butylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4[C(CH3)3]2
CAS Number:
Molecular Weight:
190.32
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.488 (lit.)

bp

106-107 °C/18 mmHg (lit.)

mp

10-11 °C (lit.)

density

0.859 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)c1cccc(c1)C(C)(C)C

InChI

1S/C14H22/c1-13(2,3)11-8-7-9-12(10-11)14(4,5)6/h7-10H,1-6H3

InChI key

ILNDSSCEZZFNGE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The solute-solvent nuclear overhauser effects (NOEs) of 1,3-di-tert-butylbenzene was studied in solvents composed of tetramethylsilane, perfluoro-tert-butyl alcohol and carbon tetrachloride.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J T Gerig
The Journal of organic chemistry, 68(13), 5244-5248 (2003-06-21)
Nuclear Overhauser effects arising from the interactions of spins of solvent molecules with spins of a solute should reveal the "exposure" of solute spins to collisions with solvent. Such intermolecular NOEs could, therefore, provide information regarding conformation or structure of
Ann-Christin Klemenz et al.
Cells, 8(7) (2019-07-13)
Metabolic characterization of human adipose tissue-derived mesenchymal stromal/stem cells (ASCs) is of importance in stem cell research. The monitoring of the cell status often requires cell destruction. An analysis of volatile organic compounds (VOCs) in the headspace above cell cultures
Natalia Drabińska et al.
Molecules (Basel, Switzerland), 24(7) (2019-04-10)
The concentration of volatile organic compounds (VOCs) can inform about the metabolic condition of the body. In the small intestine of untreated persons with celiac disease (CD), chronic inflammation can occur, leading to nutritional deficiencies, and consequently to functional impairments

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service