Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

820024

TSKgel® BioAssist® G6PW (17 µm) HPLC Columns

L x I.D. 30 cm × 7.8 mm, hydroxylated methacrylate phase

Synonym(s):

TSKgel® Size Exclusion (PW-Type) HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

Product Name

TSKgel® BioAssist® G6PW HPLC Column, phase hydroxylated methacrylate, L × I.D. 30 cm × 7.8 mm, 17 μm particle size, PEEK hardware

material

PEEK hardware

Agency

suitable for USP L38
suitable for USP L39

product line

TSKgel®

manufacturer/tradename

Tosoh 20024

parameter

≤0.5 M salt concentration
≤20% organic solvent
1.2 mL/min flow rate
10 bar max. pressure
80 °C temperature

technique(s)

HPLC: suitable

L × I.D.

30 cm × 7.8 mm

matrix

polymer particle platform

matrix active group

hydroxylated methacrylate phase

particle size

17 μm

pore size

1000 Å

operating pH

2-12

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

A TSKgel G6000PW column in PEEK column hardware, TSKgel BioAssist G6PW is for ultra-low sample adsorption during virus analysis. Use TSKgel G6000PW for the analysis of water-soluble polymers up to 8,000,000 Da molecular weight.

Application

Calibration range: up to 8,000,000 Da (polythylene glycols and oxides)

Physical form

Shipped in water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

BioAssist is a registered trademark of Tosoh Corporation
TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryan S Pralle et al.
Scientific reports, 11(1), 2839-2839 (2021-02-04)
Fatty liver syndrome is a prevalent metabolic disorder in peripartum dairy cows that unfavorably impacts lactation performance and health. Patatin-like phospholipase domain-containing protein 3 (PNPLA3) is a lipase that plays a central role in human non-alcoholic fatty liver disease etiology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service