Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

80023451

TSKgel® UP-SW Direct Connect (2 µm) HPLC Columns

L x I.D. 2 cm × 4.6 mm, diol phase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

product name

TSKgel® UP-SW Direct Connect Guard Column for P/N 80023448/9, L × I.D. 2 cm × 4.6 mm

material

stainless steel

Agency

suitable for USP L20
suitable for USP L59

product line

TSKgel®

manufacturer/tradename

Tosoh 0023451

parameter

≤0.5 M salt concentration
0-30% aqueous soluble organic solvents
10-30% temp. range

technique(s)

HPLC: suitable

L × I.D.

2 cm × 4.6 mm

matrix

silica particle platform

matrix active group

diol phase

particle size

2 μm

pore size

250 Å

operating pH

2.5-7.5

separation technique

size exclusion (SEC)

Other Notes

Related Catagories: Analytical/Chromatography, HPLC, HPLC & FPLC Columns for Biopolymer Separations, TSKgel Gel Filtration Columns for Proteins and Other Biopolymers, TSKgel Gel Filtration Columns to Analyze Purity and Aggregates, TSKgel Size Exclusion Columns

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service