Skip to Content
MilliporeSigma
All Photos(1)

Documents

68995

Supelco

Bis(2-ethylhexyl) adipate

analytical standard

Synonym(s):

Adipic acid di(2-ethylhexyl) ester, DOA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CH2CO2CH2CH(C2H5)(CH2)3CH3]2
CAS Number:
Molecular Weight:
370.57
Beilstein:
1803774
MDL number:
UNSPSC Code:
26111700
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99%

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.447 (lit.)

bp

175 °C/2 mmHg (lit.)

mp

<−70 °C (lit.)

density

0.925 g/mL at 20 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

CCCCC(CC)COC(=O)CCCCC(=O)OCC(CC)CCCC

InChI

1S/C22H42O4/c1-5-9-13-19(7-3)17-25-21(23)15-11-12-16-22(24)26-18-20(8-4)14-10-6-2/h19-20H,5-18H2,1-4H3

InChI key

SAOKZLXYCUGLFA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service