Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W447655

Sigma-Aldrich

Dioctyl adipate

≥97%, FG

Synonym(s):

Adipic acid dioctyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H42O4
CAS Number:
Molecular Weight:
370.57
FEMA Number:
4476
MDL number:
UNSPSC Code:
12352205
Flavis number:
09.951
NACRES:
NA.21
grade:
FG
Fragrance grade
food allergen:
no known allergens

grade

FG
Fragrance grade

Quality Level

Agency

follows IFRA guidelines

reg. compliance

EU 1334/2008 & 178/2002
EU Regulation 1223/2009
FDA 21 CFR 117

Assay

≥97%

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

coconut; dairy; mild

SMILES string

O(CCCCCCCC)C(=O)CCCCC(=O)OCCCCCCCC

InChI

1S/C22H42O4/c1-3-5-7-9-11-15-19-25-21(23)17-13-14-18-22(24)26-20-16-12-10-8-6-4-2/h3-20H2,1-2H3

InChI key

NEHDRDVHPTWWFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Ecotoxicity of the Adipate Plasticizers: Influence of the Structure of the Alcohol Substituent.: This study investigates the ecotoxicity of various adipate plasticizers, including dioctyl adipate, focusing on the structural influence of the alcohol substituent. It provides insights into the environmental impacts and safety profiles of these compounds (Vikhareva et al., 2021).

  • How does continuous venovenous hemofiltration theoretically expose (ex-vivo models) inpatients to diethylhexyladipate, a plasticizer of PVC medical devices: This study examines the exposure of patients to dioctyl adipate through continuous venovenous hemofiltration, emphasizing the significance of monitoring plasticizer exposure in medical settings (Bernard et al., 2020).

  • Metabolism and urinary excretion kinetics of di(2-ethylhexyl) adipate (DEHA) in four human volunteers after a single oral dose.: This research investigates the metabolic pathways and urinary excretion kinetics of dioctyl adipate in human volunteers, providing critical data for assessing human exposure and safety (Nehring et al., 2020).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service