55254-U
Supelclean™ Sulfoxide SPE Bulk Packing
pkg of 100 g
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
packaging
pkg of 100 g
Quality Level
technique(s)
solid phase extraction (SPE): suitable
matrix active group
sulfoxide phase
application(s)
food and beverages
separation technique
normal phase
General description
Supelclean Sulfoxide SPE consists of a silica-bonded sulfoxide (-SO) phase. The technology was specifically developed for the extraction of polychlorinated biphenyls (PCBs) from transformer, waste and mineral oil. PCB retention is facilitated via interaction between the SPE phase′s electrophilic sulfur atom and the π-electron cloud formed from aromatic rings inherent with PCBs. This unique SPE phase offers a simple and efficient sample prep method for identifying PCBs at quantitation limits of 0.5 ppm.
Legal Information
Supelclean is a trademark of Sigma-Aldrich Co. LLC
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service