Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55479-U

Supelco

Supel QuE QuEChERS tube

volume 15 mL, suitable for PFAS testing, pkg of 50 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104018
NACRES:
NB.11

product name

Supel QuE PSA/ENVI-Carb Tube 3, volume 15 mL, suitable for PFAS testing, pkg of 50 ea

reg. compliance

suitable for FDA C-010.02

Quality Level

packaging

pkg of 50 ea

volume

15 mL

application(s)

food and beverages

General description

The Supel QuE PSA/ENVI-Carb Tube 3, 15 mL (clean-up tube) is suitable for the testing workflow of 16 PFAS compounds in food according to regulated FDA method C_01.02. Prior to analysis by LC-MS/MS, a sample preparation is required by the "QuEChERS" method. The QuEChERS method (Quick, Easy, Cheap, Effective, Rugged, and Safe), has emerged as a sample prep technique popular e.g. in the area of matrix rich samples, like food product. In a first step, food sample is extracted with an aqueous miscible solvent (e.g., acetonitrile) in the presence of high amounts of extraction salts to induce liquid phase separation. Upon shaking and centrifugation, an aliquot of the organic phase is subjected to further cleanup step. The second step is facilitated by mixing bulk amounts of sorbent (e.g., Supelclean PSA, ENVI-Carb, MgSO4) with the extract. After sample cleanup, the mixture is centrifuged and the resulting supernatant can, depending on the sample, either be analyzed directly or can be subjected to further minor treatment before analysis.

Components

300 mg Supelclean PSA, 150 mg Supelclean, ENVI-Carb, 900 mg MgSO4

Legal Information

ENVI-Carb is a trademark of Sigma-Aldrich Co. LLC
Supel is a trademark of Sigma-Aldrich Co. LLC
Supelclean is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Discover an LC-MS/MS method for the analysis of 16 per- and polyfluoroalkyl compounds (PFAS) in milk using QuEChERS extraction technique following FDA C-010.02 guidelines.

Discover an LC-MS/MS method for the analysis of 16 per- and polyfluoroalkyl compounds (PFAS) in milk using QuEChERS extraction technique following FDA C-010.02 guidelines.

Discover an LC-MS/MS method for the analysis of 16 per- and polyfluoroalkyl compounds (PFAS) in milk using QuEChERS extraction technique following FDA C-010.02 guidelines.

Discover an LC-MS/MS method for the analysis of 16 per- and polyfluoroalkyl compounds (PFAS) in milk using QuEChERS extraction technique following FDA C-010.02 guidelines.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service