Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

54986

Coupler for Legacy HPLC Guard Column Holder

PEEK, I.D. × O.D. × Overall L 0.010 in. × 1/16 in. × 1 in.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

Product Name

Coupler for Legacy Guard Column Holder, PEEK, I.D. × O.D. × Overall L 0.010 in. × 1/16 in. × 1 in.

material

PEEK

packaging

pkg of 1 ea

technique(s)

HPLC: suitable

I.D. × O.D. × Overall L

0.010 in. × 1/16 in. × 1 in.

fitting

10-32 male (Valco-compatible)

compatibility

Also a replacement connector for stand-alone guard holder (54987)
for use with HPLC columns with 1/16", 10-32 thread end-fittings (Used with all Supelco and Astec columns, plus other brands that have the same thread dimensions.)

separation technique

chiral

General description

Finger-tight design couples HPLC columns that use 1/16" 10-32 Valco-compatible fittings with seating distance of 0.074". Also used as a replacement for PEEK coupler supplied in stand-alone guard cartridge holder, Cat. No. 54987.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service