Key Documents
54987
Holder and Coupler for Legacy HPLC Guard Columns
pkg of 1 ea, for use with non-Supelco Guard Cartridges (not TSKgel Guard Cartridges)
About This Item
UNSPSC Code:
41115700
Product Name
Holder and Coupler for Legacy Guard Columns, for use with non-Supelco guard cartridges (not TSKgel guard cartridges)
compatibility
for use with non-Supelco guard cartridges (not TSKgel guard cartridges)
General description
Finger-tight design couples HPLC columns that use 1/16" 10-32 Valco-compatible fittings with seating distance of 0.074". Also used as a replacement for PEEK coupler supplied in stand-alone guard cartridge holder, Cat. No. 54987.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)