Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51167-U

EXP® Pre-Column Filter Cartridges

0.2μm, pkg of 10 ea

Synonym(s):

TM=["EXP"] Guard Column Filters

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

packaging

pkg of 10 ea

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

EXP is a registered trademark of Optimize Technologies, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Determination of free thyroid hormones (T3 & T4) in serum by LC-MS/MS following a streamlined 1-hour BioSPME sample preparation.

Determination of free thyroid hormones (T3 & T4) in serum by LC-MS/MS following a streamlined 1-hour BioSPME sample preparation.

Determination of free thyroid hormones (T3 & T4) in serum by LC-MS/MS following a streamlined 1-hour BioSPME sample preparation.

Determination of free thyroid hormones (T3 & T4) in serum by LC-MS/MS following a streamlined 1-hour BioSPME sample preparation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service