Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51163-U

EXP® Pre-Column Filter

holder with EXP titanium hybrid ferrule, pack of 1 sets

Synonym(s):

HPLC pre-column filter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

material

(2 ferrules and 1 nut in pack)

packaging

pack of 1 sets

General description

The OPTI-SOLV® EXP® hand tight pre column filters are ideal for extreme high pressure applications. They protect columns with small particle material employing ultra high pressure techniques. The OPTI-SOLV® EXP® help extend and protect the life our your columns without sacrificing performance.
Note: cartridges not included with holder

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

EXP is a registered trademark of Optimize Technologies, Inc.
OPTI-SOLV is a registered trademark of Optimize Technologies, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Determination of free thyroid hormones (T3 & T4) in serum by LC-MS/MS following a streamlined 1-hour BioSPME sample preparation.

Determination of free thyroid hormones (T3 & T4) in serum by LC-MS/MS following a streamlined 1-hour BioSPME sample preparation.

Determination of free thyroid hormones (T3 & T4) in serum by LC-MS/MS following a streamlined 1-hour BioSPME sample preparation.

Determination of free thyroid hormones (T3 & T4) in serum by LC-MS/MS following a streamlined 1-hour BioSPME sample preparation.

Protocols

Determination of the thyroid hormones T4, T3 and rT3 from plasma using an Ascentis Express RP-Amide cartridge to trap the analytes, and a Phenyl-Hexyl column to resolve them.

BIOshell™ IgG 1000 Å C4 UHPLC Column for Improved Biomacromolecule Separations

BIOshell™ IgG 1000 Å C4 UHPLC Column for Improved Biomacromolecule Separations

BIOshell™ IgG 1000 Å C4 UHPLC Column for Improved Biomacromolecule Separations

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service