Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

502235

Supelco

ASTM® 5442 Quantitative Linearity Standard

0.01 % (w/w) each component in cyclohexane, analytical standard

Synonym(s):

ASTM® 5442 Quantitative Linearity Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

analytical standard

Agency

ASTM® D5442

CofA

current certificate can be downloaded

packaging

pkg of 6 × 1 mL

concentration

0.01 % (w/w) each component in cyclohexane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

2-30°C

Application

ASTM® 5442 Quantitative Linearity Standard is used for the analysis of petroleum waxes by gas chromatography.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Analyte

Description

    Docosane
    Dodecane
    Dotriacontane
    Eicosane
    Hexacosane
    Hexadecane
    Hexatriacontane
    Octacosane
    Octadecane
    Pentacontane
    Tetracontane
    Tetracosane
    Tetradecane
    Tetratetracontane
    Triacontane
    See All (15)

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service