Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47100

Supelco

n-Paraffin Mix C5,C6,C7,C8

(varied conc.), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

analytical standard

Agency

ASTM® D5442

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

concentration

(varied conc.)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

2-30°C

Related Categories

General description

n-Paraffin Mix C5,C6,C7,C8 may be used for determining column resolution, retention times, and response factors.
n-Paraffin Mix C5,C6,C7,C8 of analytical standard grade, is used for the analysis of petroleum waxes using gas chromatography (GC) technique.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 1 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

0.5 °F - closed cup

Flash Point(C)

-17.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service