Skip to Content
MilliporeSigma
All Photos(1)

Documents

49454-U

Supelco

Fatty Acid Ethyl Esters (FAEES), C4 - C24 Even Carbon Saturated

certified reference material, 1000 μg/mL each component in hexane

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

form

liquid

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

1000 μg/mL each component in hexane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

multi-component solution

functional group

carboxylic acid
ester

storage temp.

2-8°C

General description

Quantitative standard suitable for use as a retention index marker probe.

Check out our complete portfolio of Fatty acid methyl ester (FAME) reference material solution mixes

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Ethyl butyrate
    Ethyl hexanoate
    Ethyl octanoate
    Ethyl decanoate
    Ethyl dodecanoate
    Ethyl myristate
    Ethyl stearate
    Ethyl behenate
    Ethyl arachidate
    Ethyl palmitate
    Ethyl tetracosanoate
    See All (11)

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

5.4 °F - closed cup

Flash Point(C)

-14.8 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service