Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48495

Supelco

δ-BHC

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

CofA

current certificate can be downloaded

packaging

ampule of 50 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-30°C

SMILES string

Cl[C@@H]1[C@@H](Cl)[C@H](Cl)[C@@H](Cl)[C@H](Cl)[C@@H]1Cl

InChI

1S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H/t1-,2-,3-,4+,5-,6-

InChI key

JLYXXMFPNIAWKQ-GPIVLXJGSA-N

Application

δ-BHC may be used as a standard in determining the concentration of δ-BHC in plant extracts used in traditional chinese medicines using gas chromatography method coupled with electron capture detector (GC-ECD) or quick easy cheap effective rugged and safe multi-residue method (QuEChERS) in combination with gas chromatography coupled with electron capture detector (GC-ECD).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of pesticide residues using the Quick Easy Cheap Effective Rugged and Safe (QuEChERS) pesticide multiresidue method in traditional Chinese medicine by gas chromatography with electron capture detection
Chemosphere, 84(7), 908-912 (2011)
Residues of 18 organochlorine pesticides in 30 traditional Chinese medicines
Xue J, et al.
Chemosphere, 71(6), 1051-1055 (2008)
Jun Wu et al.
Environmental microbiology, 9(9), 2331-2340 (2007-08-10)
Commercial formulations of hexachlorocyclohexane (HCH) consist of a mixture of four isomers, alpha, beta, gamma and delta. All these four isomers are toxic and recalcitrant pollutants. Sphingobium (formerly Sphingomonas) sp. strain BHC-A is able to degrade all four HCH isomers.
Yong Qian et al.
Environmental monitoring and assessment, 116(1-3), 157-167 (2006-06-17)
The contamination of organochlorine pesticides hexachlorocyclohexane (HCH) and Dichlorodiphenyltrichloroethane (DDT) and their eco-environmental assessment in surface sediments from Lake Dongting, the second-largest freshwater lake in China, were studied. Concentrations of summation HCH (=alpha-HCH + beta-HCH + gamma-HCH +delta-HCH) were 0.21-9.59
R Calvelo Pereira et al.
Environmental pollution (Barking, Essex : 1987), 155(2), 350-358 (2007-12-26)
This study focuses on the main routes of distribution and accumulation of different hexachlorocyclohexane (HCH) isomers (mainly alpha-, beta-, gamma- and delta-HCH) in a soil-plant-air system. A field assay was carried out with two plant species, Cynara scolymus L. and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service