MilliporeSigma
  • Determination and assessment of HCHs and DDTs residues in sediments from Lake Dongting, China.
Determination and assessment of HCHs and DDTs residues in sediments from Lake Dongting, China.
Environmental monitoring and assessment (2006-06-17)
Yong Qian, Minghui Zheng, Bin Zhang, Lirong Gao, Wenbin Liu
ABSTRACT
The contamination of organochlorine pesticides hexachlorocyclohexane (HCH) and Dichlorodiphenyltrichloroethane (DDT) and their eco-environmental assessment in surface sediments from Lake Dongting, the second-largest freshwater lake in China, were studied. Concentrations of summation HCH (=alpha-HCH + beta-HCH + gamma-HCH +delta-HCH) were 0.21-9.59 ng/g dry weight and those of summation DDT = p,p'-DDD+p,p'-DDE+o,p'-DDT+p,p'-DDT) ranged from under detectable limit to 10.15 ng/g dry weight. The ratios of alpha-HCH to gamma-HCH were above 7 at most sampling sites while no or a small amount of beta-HCH were found at all sites, suggesting the degradation of HCH used in the history and possibly current use of HCH in the region. The low ratios (below 2.0 in most cases) of (p,p'-DDE+p,p'-DDD) to p,p'-DDT and high levels of individual isomers of DDT at some sites also suggested that there have still been fresh inputs of DDT into Lake Dongting. Through the comparison between concentrations of HCH and DDT residues in sediments of Lake Dongting and those from other places in China and also from the results of our eco-environmental assessment, it can be concluded that Lake Dongting is the water body with high contamination of both HCH and DDT in its sediments in comparison with other water bodies in China.
Product Description
Supelco
δ-BHC, certified reference material, 1000 μg/mL in methanol
Supelco
δ-HCH, PESTANAL®, analytical standard
Supelco
δ-BHC, PESTANAL®, analytical standard