Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47895

Supelco

Fluorobenzene solution

certified reference material, 1000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type internal

packaging

ampule of 1 mL

concentration

1000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

Fc1ccccc1

InChI

1S/C6H5F/c7-6-4-2-1-3-5-6/h1-5H

InChI key

PYLWMHQQBFSUBP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Fluorobenzene may be used as an internal standard for the quantification of volatile organic compounds in environmental samples using chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of very volatile organic compounds in environmental water by injection of a large amount of headspace gas into a gas chromatograph
Hino T, et al.
Journal of Chromatography A, 810(1-2), 141-147 (1998)
Development of a whole headspace injection method for the determination of volatile organic compounds in water
Hino T, et al.
Journal of Chromatography A, 746(1), 83-90 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service