Skip to Content
MilliporeSigma
All Photos(1)

Documents

CRM47525

Supelco

(±)-Geosmin and 2-Methylisoborneol Solution

certified reference material, TraceCERT®, 100 μg/mL each component in methanol, ampule of 1 mL

Synonym(s):

Drinking Water Odor Standards, Odor Standards

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

100 μg/mL each component in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

(±)-Geosmin and 2-Methylisoborneol are two non-toxic compounds responsible for the earthy and musty odor of water and soil. They are the secondary off-flavor metabolites produced by algae and bacteria under organic and aerobic conditions.

Application

The CRM can also be used as follows:

  • Determination of geosmin and 2-methylisoborneol (2-MIB) in recirculating aquaculture systems (RAS) by stir bar sorptive extraction (SBSE) coupled with liquid chromatography-atmospheric pressure chemical ionization-mass spectrometric (LC-APCI-MS) detection
  • Detection of geosmin and 2-methylisoborneol in 100 cyanobacterial strains using solid phase microextraction-gas chromatography-mass spectrometry (SPME GC-MS)
  • Simultaneous analysis of 51 odor-causing compounds in drinking water samples by liquid-liquid extraction (LLE) coupled with gas chromatography-triple quadrupole tandem mass spectrometry (GC-MS/MS)
  • Optimization of a headspace solid-phase microextraction-gas chromatography-mass spectrometry (HS-SPME-GC-MS) method for the quantification of off-flavor compounds in wine samples
  • Development and validation of a gas chromatography-tandem mass spectrometry (GC-MS/MS) method to measure off-flavor odorants in cork stopper samples following their headspace solid-phase microextraction using divinylbenzene/carboxen/polydimethylsiloxane (DVB/CAR/PDMS) coated fibers

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service