Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47514-U

Supelco

EPA CLP SOW OLM04 Semi-Volatile Mix solution

certified reference material, 2000 μg/mL each component in dichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Agency

US EPA/CLP SOW OLM04

description

Separate Source

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 × 1 mL (47514-U)

concentration

2000 μg/mL each component in dichloromethane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

General description

EPA CLP SOW OLM04 Semi-Volatile Mix is a multi-component solution containing certified reference materials (CRMs) such as acetophenone, atrazine, benzaldehyde, biphenyl, and ε-caprolactam.

US EPA Contract Laboratory Program (CLP) methods are analytical methods for identifying and quantifying organic compounds on the EPA′s Target Compound List (TCL) in groundwater, sediment, and soil samples at abandoned hazardous waste sites.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Acetophenone
    Atrazine
    Benzaldehyde
    Biphenyl
    ε-Caprolactam

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service