Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRM42231

Supelco

LC/MS Pesticides Calibration Mix 5

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

certified reference material
TraceCERT®

Quality Level

packaging

ampule of 1 mL
pkg of 50MG

technique(s)

LC/MS: suitable

format

multi-component solution

storage temp.

2-8°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Bendiocarb
    Benzoylprop-ethyl
    Carbofuran
    Demeton-S-methyl-sulfon
    Diethofencarb
    Disulfoton-sulfone
    Ethiofencarb
    Fenoxycarb
    Fenthion-sulfone
    Fenthionsulfoxide
    Flonicamid
    Furathiocarb
    Methiocarb
    Omethoate
    Pencycuron
    Promecarb
    Propoxur
    Carbaryl
    Spiromesifen
    Sulfentrazone
    Tebufenozide
    Tepraloxydim
    Thiabendazole
    Thiacloprid
    Thiametoxam
    Thiodicarb
    Thiofanox
    Thiofanox-sulfoxide
    Tralkoxydim
    Trifloxystrobin
    Zoxamide
    See All (31)

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service