Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47102

Supelco

n-Paraffin Mix C10,C12,C14,C16

certified reference material, (varied conc.)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

Quality Level

Agency

ASTM® D5442

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

concentration

(varied conc.)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

2-30°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany
ASTM is a registered trademark of American Society for Testing and Materials

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

114.8 °F

Flash Point(C)

46 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service