Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44808

Supelco

Aroclor 1254 solution

certified reference material, 1000 μg/mL in isooctane

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
77101502

grade

certified reference material
TraceCERT®

Quality Level

Agency

EPA 8082

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

1000 μg/mL in isooctane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

InChI

1S/C12H5Cl5/c13-8-3-1-2-6(10(8)15)7-4-5-9(14)12(17)11(7)16/h1-5H

InChI key

AUGNBQPSMWGAJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Piekoszewski et al.
Folia medica Cracoviensia, 31(3), 201-207 (1990-01-01)
The pharmacokinetics of antipyrine in a dose of 20 mg/kg i.v. and cytochrome P-450 level in liver, were studied in control and pretreated rats with 1, 5, 25 ppm polychlorinated biphenyls in food for up to 120 days. The objective
Cyril Rigaud et al.
Aquatic toxicology (Amsterdam, Netherlands), 149, 133-144 (2014-03-13)
The 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD) toxic equivalent quantity (TCDD-TEQ) approach was used successfully to predict lethal embryotoxicity in salmonids, but its applicability to sublethal effects of mixtures of organohalogenated compounds in other fish species is poorly known. The sublethal toxicity of two
Qiuli Shan et al.
Toxicology and applied pharmacology, 276(2), 136-146 (2014-03-04)
2,3,7,8-Tetrachlorodibenzo-p-dioxin (TCDD) and polychlorinated biphenyls (PCBs) are persistent organic pollutants found as complex mixtures in the environment throughout the world. Therefore, humans are ubiquitously and simultaneously exposed to TCDD and PCBs. TCDD and PCBs alone have been linked to atherosclerosis.
D L Arnold et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 31(1), 42-55 (1996-05-01)
A total of 80 menstruating rhesus monkeys (Macaca mulatta) were equally and randomly divided among groups receiving 0, 5, 20, 40, or 80 mu g of Aroclor 1254/kg body weight/day during a 6-year toxicological-reproduction study. During the first 3 years
Javier Esteban et al.
Toxicology letters, 229(1), 41-51 (2014-06-03)
Polychlorinated biphenyls (PCBs) induce a broad spectrum of biochemical and toxic effects in mammals including alterations of the vital retinoid (vitamin A) system. The aim of this study was to characterize alterations of tissue retinoid levels in rat offspring and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service