Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

38273

Supelco

Prothioconazole

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

2-[2-(1-Chlorocyclopropyl)-3-(2-chlorophenyl)-2-hydroxypropyl]-1,2-dihydro-3H-1,2,4-triazole-3-thione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15Cl2N3OS
CAS Number:
Molecular Weight:
344.26
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

2-8°C

SMILES string

OC(CN1N=CNC1=S)(Cc2ccccc2Cl)C3(Cl)CC3

InChI

1S/C14H15Cl2N3OS/c15-11-4-2-1-3-10(11)7-14(20,13(16)5-6-13)8-19-12(21)17-9-18-19/h1-4,9,20H,5-8H2,(H,17,18,21)

InChI key

MNHVNIJQQRJYDH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Application

  • Quaternized chitosan-based organic-inorganic nanohybrid nanoparticles loaded with prothioconazole for efficient management of fungal diseases with minimal environmental impact: This study highlights the development of an advanced formulation of prothioconazole using quaternized chitosan-based nanoparticles, focusing on efficient fungal disease management in agricultural settings. This application demonstrates prothioconazole′s role in sustainable agriculture by reducing the environmental impact of fungicide use, crucial for researchers in agricultural biotechnology and environmental science (Niu et al., 2024).
  • Effect of novel botanical synergist on the effectiveness and residue behavior of prothioconazole in wheat field: Research demonstrates the enhancement of prothioconazole′s fungicidal efficacy with a novel botanical synergist. This study is pivotal for those investigating the dynamic interactions of chemical and botanical fungicides, aiming to optimize crop protection while monitoring residue levels for food safety (Wu et al., 2023).
  • Exploring the Potential Mechanism of Prothioconazole Resistance in Fusarium graminearum in China: This research addresses the growing concern of fungicide resistance, particularly investigating the mechanisms behind Fusarium graminearum′s resistance to prothioconazole. The findings are essential for biotechnologists and plant pathologists working to develop new strategies to overcome resistance in crop pathogens (Zhou et al., 2023).
  • Establishing the extent of pesticide contamination in Irish agricultural soils: This study utilizes prothioconazole as a marker to assess the extent of pesticide contamination in soils. It is critical for environmental researchers and regulatory bodies aiming to understand the environmental impact of prolonged fungicide use and to develop guidelines for sustainable farming practices (Vickneswaran et al., 2023).

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service