Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

2879905-U

Supelco

Inlet Liner, Split/Splitless Type, Single Taper FocusLiner Design (wool packed)

pkg of 5 ea

Synonym(s):

GC accessory, glass wool, inlet liner

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115719
NACRES:
SB.54

packaging

pkg of 5 ea

L × O.D. × I.D.

78.5 mm × 6.3 mm × 4.0 mm

Looking for similar products? Visit Product Comparison Guide

General description

Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits Agilent (5890, 6890, and 7890)
  • Fits Finnigan (9001GCQ)
  • Fits Varian (CP-1177 Injector)

Application


  • Split/Splitless Inlets in Gas Chromatography: What′s Up with All Those Different Glass Inlet Liners: This article provides an in-depth review of the various configurations of glass inlet liners used in split and splitless inlets for gas chromatography. The Single Taper FocusLiner Design (wool packed) is highlighted for its effectiveness in enhancing sample vaporization and reducing analyte loss, making it ideal for high-precision analytical applications in complex sample matrices (Snow, 2022).

Legal Information

FocusLiner is a trademark of SGE Analytical Science Pty Ltd.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Caterina Agrimonti et al.
International journal of food microbiology, 305, 108246-108246 (2019-06-27)
Cellulosic pads, amended with emulsions containing essential oils of thyme and oregano, exhibited antimicrobial activity against the psychrophilic microbiota of minced beef. In addition, the pads were active against specific meat bacterial species (Pseudomonas putida, Pseudomonas fragi, Pseudomonas fluorescens, Enterococcus

Articles

GC-MS method detects nitrosamines in Valsartan tablets, meeting US FDA guidelines for pharmaceutical quality control.

GC-MS method detects nitrosamines in Valsartan tablets, meeting US FDA guidelines for pharmaceutical quality control.

GC-MS method detects nitrosamines in Valsartan tablets, meeting US FDA guidelines for pharmaceutical quality control.

GC-MS method detects nitrosamines in Valsartan tablets, meeting US FDA guidelines for pharmaceutical quality control.

See All

Protocols

Complete workflow for the comprehensive analysis of terpenes in cannabis

Complete workflow for the comprehensive analysis of terpenes in cannabis

Complete workflow for the comprehensive analysis of terpenes in cannabis

Complete workflow for the comprehensive analysis of terpenes in cannabis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service