Key Documents
24843
Supelco
Mini-Buck Flow Calibrator
model, M-5, flow rate 1-6000 mL/min
About This Item
UNSPSC Code:
41104008
feature
model M-5
flow rate measuring range
1-6000 mL/min
technique(s)
active air sampling: suitable
application(s)
air monitoring
environmental
industrial hygiene
General description
Battery charger not included, order separately.
Application
Mini-buck flow calibrator is a gas flow calibrator, which may be used for the development and evaluation of a calibration gas generator based on the injection method for the analysis of volatile organic compounds in air.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Development and evaluation of a calibration gas generator for the analysis of volatile organic compounds in air based on the injection method.
Rosenberg E, et al.
Fresenius Journal of Analytical Chemistry, 371(6), 798-805 (2001)
Protocols
Simplify your thermal desorption-based terpenes sample collection in air. Learn more about their profiling, odor measurement, industrial hygiene, and environmental emissions.
Simplify your thermal desorption-based terpenes sample collection in air. Learn more about their profiling, odor measurement, industrial hygiene, and environmental emissions.
Simplify your thermal desorption-based terpenes sample collection in air. Learn more about their profiling, odor measurement, industrial hygiene, and environmental emissions.
Simplify your thermal desorption-based terpenes sample collection in air. Learn more about their profiling, odor measurement, industrial hygiene, and environmental emissions.