Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24518

Supelco

Supelpure HC Hydrocarbon Trap

volume 750 cc, 1/4 in. fitting

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41115700

volume

750 cc

Quality Level

fitting

1/4 in. fitting

Looking for similar products? Visit Product Comparison Guide

General description

Activated charcoal adsorbs hydrocarbons and other contaminants from carrier gases, air, and hydrogen. Operates efficiently for approximately six months when total hydrocarbons in the incoming gas average 10 ppm.

Dimensions:
  • 120 cc Trap: 111/8in. (28.2 cm) long x 13/8 in. (35 mm) diameter
  • 750 cc Trap: 161/2 in. (41.9 cm) long x 25/16in. (59 mm) diameter
  • S-Trap: 71/2 in. (19 cm) long x 59/16 in. (14.1 cm) wide x 1/2 in. (13 mm) diameter [total bed length = 193/4 in. (50.2 cm); the extended bed length ensures prolonged contact between the gas and the adsorbent and provides greater working capacity]
Hydrocarbon traps are basically used in GC for carrier gas purification, as the name suggests they are primarily used for trapping hydrocarbons. Some hydrocarbon traps, like the Supelpure HC Hydrocarbon Trap, contain a carbon molecular sieve that also restricts the elution of moisture and oxygen. Surges of moisture and oxygen that enter the gas line during the installation of a new gas cylinder enter the Supelpure HC Hydrocarbon Trap and are released slowly, diluted by the clean carrier gas from the new cylinder. It particularly removes C3 and other higher hydrocarbons.

Legal Information

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service