Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

503215

Supelco

Carbon Dioxide Trap

volume 250 cc, 1/4 in. fitting

Synonym(s):

CO2 TRAP 250CC, 1/4IN FITTINGS

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12000000

volume

250 cc

fitting

1/4 in. fitting

Looking for similar products? Visit Product Comparison Guide

General description

Sodium hydroxide nonfibrous silicate material is universally accepted for its high absorptive capacity and indicating properties for carbon dioxide. Typically, this material will absorb 20-30% of its weight in carbon dioxide before replacement of the saturated material is required. As carbon dioxide is absorbed, the greenish-brown material turns white because of the formation of sodium carbonate. The trap body is constructed of borosilicate glass, and the fittings are nickel-plated and have sintered stainless steel frits.

A moisture trap should be installed downstream from this unit to absorb the moisture that is produced.

Note: Exercise extreme caution when refilling the trap due to the caustic nature of the absorbent.

Dimensions:
  • 100 cc Traps: 123/4 in. (32.4 cm) long x 13/4 in. (44 mm) diameter
  • 250 cc Traps: 171/2 in. (44.4 cm) long x 2 in. (51 mm) diameter

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service