Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

23900-U

Supelco

OMI® Purifier Tube

OMI-1 Purifier Tube

Synonym(s):

OMI®-1 INDICATOR TUBE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
SB.54

description

includes 2 ferrules

Looking for similar products? Visit Product Comparison Guide

General description

The OMI is a polishing purifier that removes many contaminants that other upstream purifiers miss. It will simultaneously and irreversibly remove moisture, oxygen, carbon monoxide, carbon dioxide, most sulfur compounds, most halogen compounds, alcohols, and phenols to less than 10 ppb. It is recommended that an OMI be installed in carrier gas streams just upstream of every GC. It consists of two components:
  • A re-useable tube holder that is installed into the gas delivery system. The use of polycarbonate provides see-through capability along with safety.
  • A purifier tube containing an indicating resin (changes color from black to brown when exposed to as little as 1 ppm of moisture or oxygen). The use of glass provides see-through capability and prevents diffusion of room contaminants into the gas stream.

Spent purifier tubes are easily replaced. Simply unscrew the end assembly from the tube holder and replace it with a new purifier tube. The design prevents room air from entering the new tube during installation (protective foil covers on each end are only pierced as the end assembly is screwed back onto the tube holder to complete installation).

Note: This item requires a re-useable tube holder (purchased separately).

Footnote

For use with users who have an OMI-1 installation kit (no longer available).

Legal Information

OMI is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Water-react 1

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service