Skip to Content
MilliporeSigma

1.13306

Bactident Coagulase

For the detection of the enzyme coagulase developed by Staphylococcus aureus

Synonym(s):

Test on Staphylococcus aureus, Test on Coagulase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.84
UNSPSC Code:
41116130
Material:
tube
Packaging:
pack of 6 vials with 3 mL volume
Suitability:
Staphylococcus aureus, Staphylococcus spp. (spp.)
Feature:
ready-to-use
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

tube

form

solid

feature

ready-to-use

packaging

pack of 6 vials with 3 mL volume

manufacturer/tradename

Bactident

storage condition

(Store dry and tightly closed)

technique(s)

microbe id | specific enzyme detection: suitable

color

beige

application(s)

food and beverages

storage temp.

2-8°C

suitability

Staphylococcus aureus, Staphylococcus spp. (spp.)

General description

Bactident Coagulase reagent enables reliable detection of the enzyme coagulase developed by Staphylococcus aureus. Coagulase is an enzyme with the ability to coagulate plasma. Staphylococcus aureus forms two types of coagulases; (i) the free coagulase is an extracellular enzyme, and (ii) the bound coagulase is localized on the surface of the cell wall. The tube test can detect both enzymes, while the slide test can detect only the bound coagulase.

Application

An overnight broth culture of Staphylococcus aureus is incubated with rehydrated EDTA-rabbit plasma, which is contained in Bactident Coagulase. The coagulase test is positive if more than three-quarters of the tube contents form a coherent clot.

Analysis Note

Coagulase
- Staphylococcus aureus ATCC 25923 (WDCM 00034):
+
- Staphylococcus epidermidis ATCC 12228 (WDCM 00036):-
- Negative control:-

Legal Information

BACTIDENT is a trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library