Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

20349

Supelco

ORBO 47 Amberlite XAD®-7 specially treated, 100/50 mg

W,W,W separators, O.D. × L 6 mm × 90 mm, pkg of 50 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131900

material

W,W,W separators

Agency

NIOSH 2546
OSHA 32,110

product line

Amberlite
ORBO

composition

Bed A, 100 mg
Bed B, 50 mg

packaging

pkg of 50 ea

manufacturer/tradename

ORBO 47

technique(s)

active air sampling: suitable

O.D. × L

6 mm × 90 mm

matrix

XAD-7 (specially treated)

application(s)

air monitoring
environmental
industrial hygiene

General description

ORBO sorbent tubes contain two beds of the same selective adsorbent separated by glass wool or foam, for gas and vapor sampling. The dual-layer or two-bed construction of the tube allows for any sample breakthrough to be captured in the smaller back-up bed.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.
ORBO is a trademark of Sigma-Aldrich Co. LLC
XAD is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liang Yang et al.
Cell reports, 24(3), 585-593 (2018-07-19)
In response to blue light, cryptochromes photoexcite and interact with signal partners to transduce signal almost synchronously in plants. The detailed mechanism of CRY-mediated light signaling remains unclear: the photobiochemical reactions of cryptochrome are transient and synchronous, thus making the monitoring and
Yi Li et al.
Cell chemical biology, 27(7), 858-865 (2020-05-30)
The engineered ascorbate peroxidase (APEX) is a powerful tool for the proximity-dependent labeling of proteins and RNAs in live cells. Although widely use in mammalian cells, APEX applications in microorganisms have been hampered by the poor labeling efficiency of its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service