Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05346

Supelco

Decanoic acid-1,2,3,4-13C4

analytical standard

Synonym(s):

Capric acid-1,2,3,4-13C4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C4C6H20O2
CAS Number:
Molecular Weight:
176.24
UNSPSC Code:
12352106
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

extent of labeling

≥99%

storage temp.

2-8°C

SMILES string

CCCCCC[13CH2][13CH2][13CH2][13C](O)=O
CCCCCC[13CH2][13CH2][13CH2][13C](O)=O

InChI

1S/C10H20O2/c1-2-3-4-5-6-7-8-9-10(11)12/h2-9H2,1H3,(H,11,12)/i7+1,8+1,9+1,10+1

InChI key

GHVNFZFCNZKVNT-HZKSPUCSSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service