Skip to Content
MilliporeSigma
All Photos(8)

Key Documents

ZRB2223

Sigma-Aldrich

Anti-CA9 Antibody, clone 1B6 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

CA-IX, CAIX, Carbonate dehydratase IX, Carbonic anhydrase 9, Carbonic anhydrase IX, EC:4.2.1.1, Membrane antigen MN, P54/58N, RCC-associated antigen G250, Renal cell carcinoma-associated antigen G250, pMW1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1B6, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

purified by

using Protein A

species reactivity

human, rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal cytoplasmic domain

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... CA9(768)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1B6 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Carbonic anhydrase 9 (CA9). It targets an epitope within 24 amino acids from the cytoplasmic domain in the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 24 amino acids from the C-terminal, cytoplasmic domain of human Carbonic anhydrase 9 (CA().

Application

Quality Control Testing

Evaluated by Western Blotting in Rat colon tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected CA9 in Rat colon tissue lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected CA9 in Human stomach tissue lysate.

Affinity Binding Assay: A representative lot of this antibody bound CA9 with a KD of 9.2 x 10-8 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected CA9 in Human stomach tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected CA9 in A431 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Carbonic anhydrase 9 (UniProt: Q16790; also known as EC:4.2.1.1, Carbonate dehydratase IX, Carbonic anhydrase IX, CA-IX, CAIX, Membrane antigen MN, P54/58N, Renal cell carcinoma-associated antigen G250, RCC-associated antigen G250, pMW1) is encoded by the CA9 (also known as G250, MN) gene (Gene ID: 768) in human. Six evolutionary distinct classes of carbonic anhydrases (CA) have been described (α, β, γ, δ, ζ and η). However, only the α-class is expressed in mammals. Sixteen different α-CA isozymes have been described that vary in their catalytic activity and subcellular localization. Out of the twelve catalytically active forms five are cytosolic, four are membrane bound with their catalytic domain facing the extracellular space, two are mitochondrial, and one is secreted isoform. They catalyze the conversion of CO2 to the bicarbonate iOn and protons to mediate pH and CO2 homeostasis. CA9, a membrane bound isoform, is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-37), which is subsequently cleaved off to produce the mature form that contains an extracellular domain (aa 38-414), a transmembrane domain (aa 415-435), and a short cytoplasmic domain (aa 436-459). It′s α-carbonic anhydrase domain is localized in amino acids 139-390. In addition, it features a proteoglycan-like domain (PG) in its N-terminal region (aa 38-112) that is unique to CA9 and contributes to the assembly of focal adhesion contacts during cell migration and may function as a proton buffer to support its catalytic activity. CA9 expression in normal healthy tissues is restricted to epithelial cells in the stomach and gut but is strongly upregulated in many tumor tissues. Its expression is under the control of hypoxia-inducible factor 1 (HIF-1). CA9 expression is observed in hypoxic and mild hypoxic  regions. Its expression is reported in a breast and colorectal cancer, glioblastoma, lung cancer, and cervical squamous cell carcinomas, and is usually linked to poor prognosis. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, Affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Becker, HM. (2020). Br. J. Cancer 122(2); 157-167; Hilvo, M., et al. (2008). J. Biol. Chem. 283(410; 27799-27809).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

Affinity is a trademark of Mine Safety Appliances Co.
ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany
iOn is a trademark of 3DXTech

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service