Skip to Content
MilliporeSigma
All Photos(13)

Key Documents

ZRB1943

Sigma-Aldrich

Anti-Notch1 Antibody, clone 1A32 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1A32, recombinant monoclonal

description

1A32 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 272.51 kDa
observed mol wt ~125 kDa

species reactivity

human

species reactivity (predicted by homology)

mouse, rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1A32 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Notch1. It targets an epitope within 13 amino acids from the extracellular domain in the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 13 amino acids from the extracellular domain within the C-terminal half of human Notch1.

Application

Quality Control Testing

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Notch1 in HEK293 cell lysate.

Tested applications

Western Blotting Analysis: A 1:1000 dilution from a representative lot detected Notch1 from wild-type HEK293 cells, but not from Notch1 knockout cells.

Immunocytochemistry Analysis: A 1:200 dilution from a representative lot detected Notch1 in HeLa and A431 cells.

Affinity Binding Assay: A representative lot of this antibody bound Notch1 peptide with a KD of 1.0 x 10-12 in an affinity binding assay.


Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Notch1, clone 1A32 ZooMAb®, Cat. No. ZRB1943, is a recombinant Rabbit monoclonal antibody that detects Notch1 and is tested for use in Affinity Binding Assay, Immunocytochemistry, and Western Blotting.

Target description

Neurogenic locus notch homolog protein 1 (UniProt: P46531; also known as Notch 1, hN1, Translocation-associated notch protein TAN-1) is encoded by the NOTCH1 (also known as TAN1) gene (Gene ID: 4851) in human. Notch1 is a single-pass type I membrane protein that is synthesized with a signal peptide (aa 1-18), which is subsequently cleaved off to generate the mature form that contains a long extracellular domain (aa 19-1735), a transmembrane domain (aa 1736-1756), and a cytoplasmic domain (aa 1757-2555). It is expressed in most fetal tissues and is found in abundance in spleen, brain stem, and lung tissue. It is also found in most adult lymphoid tissues. Notch1 is synthesized in the endoplasmic reticulum in an inactive form and is proteolytically cleaved by a furin-like convertase in the trans-Golgi network before it reaches the plasma membrane to yield an active, ligand-accessible form. Cleavage results in a C-terminal fragment N(TM) and a N-terminal fragment N(EC). Following ligand binding, it is cleaved by ADAM17 to yield a membrane-associated intermediate fragment called notch extracellular truncation. Following endocytosis, this fragment is then cleaved by one of the catalytic subunits of g-secretase to release a Notch-derived peptide containing the intracellular domain (NICD) from the membrane. Notch 1 functions as a receptor for membrane-bound ligands Jagged-1 (JAG1), Jagged-2 (JAG2) and Delta-1 (DLL1) to regulate cell-fate determination. Upon ligand activation through the released NICD, it forms a transcriptional activator complex with RBPJ/RBPSUH and activates genes of the enhancer of split locus. Notch 1 is reported to be involved in angiogenesis and negatively regulates endothelial cell proliferation and migration and angiogenic sprouting. It is also involved in the maturation of both CD4+ and CD8+ cells in the thymus. Notch1 represses neuronal and myogenic differentiation and plays a role in post-implantation development, including mesoderm development, somite formation and neurogenesis. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

600 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service