Skip to Content
MilliporeSigma
All Photos(8)

Key Documents

ZRB2006

Sigma-Aldrich

Anti-phospho-p38 (T180/Y182) Antibody, clone 2B7 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

EC:2.7.11.24, MAP kinase 13, MAP kinase p38 delta, MAPK 13, Mitogen-activated protein kinase 13, Mitogen-activated protein kinase p38 delta, Stress-activated protein kinase 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2B7, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
inhibition assay: suitable
western blot: suitable

isotype

IgG

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2B7 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects p38 MAP kinase phosphorylated on threonine 180 and tyrosine 182.

Immunogen

KLH-conjugated linear peptide corresponding to 14 amino acids surrounding phosphothreonine 180 and phosphotyrosine 182 of human p38 MAP kinase delta.

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from HEK293 cells treated with Anisomycin.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected phospho-p38 MAPK (Thr180/Tyr182) in lysate from overnight serum starved HEK293 cells treated with Anisomycin (5 g/mL; 30 min.)

Tested Applications

Peptide Inhibition Assay:: Target band detection in lysate from HEK293 cells treated with Anisomycin was prevented by pre-blocking of a representative lot with the immunogen phosphopeptide, but not the corresponding non-phosphopeptide.

Flow Cytometry Analysis: 0.1 µg from a representative lot detected phospho-p38 MAPK (Thr180/Tyr182) in one million HEK293 cells treated with Anisomycin.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected phospho-p38 MAPK (Thr180/Tyr182) in Human colon cancer tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Mitogen-activated protein kinase 13 (UniProt: O15264; also known as EC:2.7.11.24, MAP kinase 13, MAPK 13, Mitogen-activated protein kinase p38 Delta, MAP kinase p38 Delta, Stress-activated protein kinase 4) is encoded by the MAPK13 (also known as PRKM13, SAPK4) gene (Gene ID: 5603) in human. p38 is a serine/threonine kinase that is an essential component of the MAP kinase signal transduction pathway. It plays an important role in the cascades of cellular responses evoked by extracellular stimuli such as pro-inflammatory cytokines or physical stress leading to direct activation of transcription factors such as ELK1 and ATF2. Four splice variants of the p38 family have been identified and are known as p38α, p38β, p38γ, and p38δ. Of these, p38α and p38β are ubiquitously expressed while p38γ and p38δ are differentially expressed depending on tissue type. p38δ expression is observed in testes, pancreas, small intestine, lung, and kidney. It is also abundant in macrophages and is also detected in neutrophils, CD4+ T-cells, and endothelial cells. It phosphorylates a broad range of downstream proteins. Its protein kinase domain is localized in amino acids 25-308. It is fully activated following Dual® phosphorylation on threonine 180 and tyrosine 182 by MKK3, MKK4, MKK6, and MKK7. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, Affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Ozawa, S., et al. (2010). Biochem. Biophys. Res. Commun. 396(4); 1060-1064; Zarubin, T., and Han, J. (2005). Cell Res. 15(1); 11-18; Jiang, Y., et al. (1997). J. Biol. Chem. 272(48); 30122-30128).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

Affinity is a trademark of Mine Safety Appliances Co.
Delta is a trademark of Heathrow Scientific LLC
Dual is a registered trademark of Ciba-Geigy AG
ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service