Skip to Content
MilliporeSigma
All Photos(16)

Key Documents

ZRB1173

Sigma-Aldrich

Anti-phospho JNK (Thr183, Tyr185) Antibody, clone 3F7, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3F7, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 48.29 kDa

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
inhibition assay: suitable (peptide)
western blot: suitable (peptide)

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

phosphorylation (pThr183/pTyr185)

Gene Information

human ... MAPK8(5599)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 3F7 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects JNK1/2 phosphorylated on threonine 183 and tyrosine 185.

Immunogen

KLH-conjugated linear peptide corresponding to 10 amino acids surrounding phosphothreonine 183 and phosphotyrosine 185 from human JNK/SAPK.

Application

Anti-phospho JNK (Thr183, Tyr185), clone 3F7 ZooMAb , Cat. No. ZRB1173, is a highly specific recombinant rabbit monoclonal antibody that detects JNK/SAPK phosphorylated on Thr183 and Tyr185 and has been tested for use in Immunocytochemistry, Immunohistochemistry, Peptide Inhibition Assay, and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected phospho JNK (Thr183, Tyr185) in NIH3T3 cells treated with Anisomyocin.

Immunohistochemistry (Paraffin) Analysis: A 1:100-1,000 dilution from a representative lot detected phospho JNK (Thr183, Tyr185) in human colon cancer and human ovarian cancer tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected phospho JNK (Thr183, Tyr185) in HeLa cells treated with Anisomycin treatment.

Peptide Inhibition Analysis: A 1:1,000 dilution from a representative lot was used with NIH3T3 cells treated with Anisomyocin ((25 ug/ml for 30 min following overnight serum starvation) for peptide block analysis.

Note: Actual optimal working dilutions must be determined by the end user as specimens and experimental conditions may vary.

Target description

Mitogen-activated protein kinase 8/9 (UniProt: P45983/P45984; also known as MAP kinase 8/9, MAPK8/9, Stress-activated protein kinase 1c/1a, SAPK1c/SAPK1a, Stress-activated protein kinase JNK1/ JNK2, c-Jun N-terminal kinase 1/2) is encoded by the MAPK8/9 (also known as JNK1/JNK2, PRKM8/9, SAPK1/2, SAPK1C/1A) gene (Gene ID: 5599/ 5601) in human. SAPK/JNK is a serine/threonine-protein kinase that is involved in various biological processes, including cell proliferation, differentiation, migration, transformation, and programmed cell death. It is activated in response to extracellular stimuli such as proinflammatory cytokines or physical stress. Both JNK1 and JNK2 contain a TXY motif (aa 183-185) that contains the threonine and tyrosine residues whose phosphorylation activates these kinases. Protein kinase domain of SAPK/JNK is localized to amino acids 26-321. Phosphorylation and activation of SAPK/JNK is achieved by two dual specificity kinases, MAP2K7 and MAP2K4. Following their activation SAPK/JNK phosphorylate several transcription factors, primarily components of AP-1 such as JUN, JDP2, and ATF2 and thus regulates AP-1 transcriptional activity. It promotes apoptotic cell death in stressed cells by phosphorylating key regulatory factors including p53/TP53 and Yes-associates protein YAP1. The immunogen sequence used to generate this clone is conserved in both JNK1 and JNK2. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose. Normal appearance is a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rafia S Al-Lamki et al.
The American journal of pathology, 192(4), 722-736 (2022-01-23)
Similar to the behavior of inflamed tubular epithelial cells, clear cell renal cell carcinoma (ccRCC) cells express death receptor 3 (DR3 or TNFSFR25) in situ, and expression increases with tumor grade. Surprisingly, E-selectin, which can be induced in endothelial cells
Jie Liu et al.
Molecular medicine reports, 24(3) (2021-07-20)
Previous studies have identified microRNA (miRNA/miR)‑3613‑3p as a heat stress (HS)‑related miRNA in endothelial cells that can lead to apoptosis. However, the mechanism underlying the miR‑3613‑3p‑mediated apoptosis of HS‑exposed endothelial cells remains unclear. In the present study, western blot analysis
Chengliang Yuan et al.
Journal of biochemical and molecular toxicology, 36(1), e22943-e22943 (2021-11-02)
Eriocitrin, a lemons flavanone, exhibits several biological properties, antiproliferative, and proapoptotic effects. However, its molecular mechanical action is not entirely clarified. Oxidative stress causes abnormal stimulation of signal transducer and activator of transcription 3 (STAT3) and c-Jun NH2-terminal kinase (JNK), p38

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service