Skip to Content
MilliporeSigma
All Photos(10)

Key Documents

ZRB1989

Sigma-Aldrich

Anti-HLA-G Antibody, clone 1C18 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

HLA class I histocompatibility antigen, alpha chain G;HLA G antigen;MHC class I antigen G

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1C18, recombinant monoclonal

description

1C18 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 38.22 kDa
observed mol wt ~40 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... HLA-G(3135)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1C18 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects HLA class I histocompatibility antigen, alpha chain G (HLA-G). It targets an epitope within 18 amino acids from the extracellular domain within the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 18 amino acids from the extracellular domain within the N-terminal half of human HLA class I histocompatibility antigen, alpha chain G (HLA-G).

Application

Quality Control Testing

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected HLA-G in Jurkat cell lysate.

Tested applications

Affinity Binding Assay: A representative lot of this antibody bound HLA-G peptide with a KD of 2.4 x 10-7 in an affinity binding assay.

Flow Cytometry Analysis: 1 μg from a representative lot detected HLA-G in one million JEG3 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected HLA-G in human placenta tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected HLA-G in Jurkat cell lysate.

Target description

HLA class I histocompatibility antigen, alpha chain G (UniProt: P17693; also known as HLA G antigen, MHC class I antigen G) is encoded by the HLA-G (also known as HLA-6.0, HLAG) gene (Gene ID: 3135) in human. HLA-G is a class I histocompatibility antigen, but unlike the classical class I major histocompatibility complex, it displays limited polymorphism. Seven different isoforms of HLA-G have been described that are produced by alternative splicing. With the exception of isoform 1, other isoforms lack a portion of their extracellular domain. HLA-G is a single-pass type I membrane protein that is synthesized with a signal peptide (aa 1-24), which is subsequently cleaved off to generate the mature form that contains an extracellular domain (aa 25-308), a transmembrane domain (aa 309-332), and a short cytoplasmic tail (aa 333-338). It is also present on endoplasmic reticulum and endosomes. HLA-G expression is observed in adult eye and in immune cell subsets including monocytes, myeloid and plasmacytoid dendritic cells, and regulatory T cells. It is also expressed abundantly and specifically on extravillous trophoblasts (EVT) in physiological conditions. Soluble HLA-G is also found in maternal blood, amniotic fluid, and cord blood and participates in maternal-fetal immune tolerance by keeping in check the effector functions of NK, CD8+ T cells, and B cells. Its expression is detected as early as the morula stage. HLA-G gene polymorphisms and reduced levels of soluble HLA-G have been linked to failure of embryo implantation, recurrent spontaneous abortions, and placental abruption. HLA-G is considered as a critical marker of immunotolerance in cancer cell immune evasion and is strongly associated with disease progress and prognosis for cancer patients. Its expression and isoform profile varies significantly among tumor types and between primary tumor and metastases. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Xu, X., et al. (2020). Front. Immunol. 11; 592010; Lin, A., and Yan, W-H. (2018). Front. Immunol. 9; 2164).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service