Skip to Content
MilliporeSigma

CBL1360

Anti-CD117 Antibody, clone ACK2

clone ACK2, Chemicon®, from rat

Synonym(s):

c-kit, SCF Receptor

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
ACK2, monoclonal
Application:
FACS, IHC, IP, WB
Citations:
8
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rat

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

ACK2, monoclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

input

sample type hematopoietic stem cell(s)
sample type mesenchymal stem cell(s)

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KIT(3815)

General description

145 kDa
This 145 kDa molecule is a member of the tyrosine kinase receptor family and is expressed by a majority of hematopoietic progenitor cells. The interaction of the mouse c-kit receptor and steel factor promotes proliferation and differentiation of hematopoietic progenitor cells. CD117 is also expressed by mast cells, and plays a role in signaling and activation of these cells.

Application

Detect CD117 using this Anti-CD117 Antibody, clone ACK2 validated for use in FC, IH, IP, FUNC & WB.
Flow Cytometry: 0.2 μg per 10e6 cells

Immunoprecipitation

Immunohistochemistry (Frozen)

Western blot

Blocking Ligand Binding

Optimal working dilutions must be determined by the end user.
Research Category
Stem Cell Research
Research Sub Category
Hematopoietic Stem Cells

Mesenchymal Stem Cells

Biochem/physiol Actions

Specifically recognizes mouse CD117, known as c-kit receptor, stem cell factor receptor and steel factor receptor.

Physical form

Format: Purified
Protein A purified
The monoclonal is presented in phosphate buffered saline containing 10mM sodium azide.

Preparation Note

For use within 1 month of purchase store at +4°C, for long term storage aliquot antibody into small volumes and store at -20°C.

Analysis Note

Control
Skin tissue, normal breast epithelium, melanocytes, mast cells and glial cells

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: CBL1360P

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library