Skip to Content
MilliporeSigma
All Photos(9)

Key Documents

ZRB1936

Sigma-Aldrich

Anti-RUNX1 Antibody, clone 1C18 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Runt-related transcription factor 1;Acute myeloid leukemia 1 protein;Core-binding factor subunit alpha-2;CBF-alpha-2, Oncogene AML-1;Polyomavirus enhancer-binding protein 2 alpha B subunit;PEA2-alpha B;PEBP2-alpha B;SL3-3 enhancer factor 1 alpha B subunit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
conjugate:
unconjugated
application:
ICC
WB
affinity binding assay
clone:
1C18, recombinant monoclonal
species reactivity:
human, rat, mouse
citations:
technique(s):
affinity binding assay: suitable
immunocytochemistry: suitable
western blot: suitable

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1C18, recombinant monoclonal

description

1C18 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 48.74 kDa
observed mol wt ~52 kDa

purified by

using Protein A

species reactivity

human, rat, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... RUNX1(861)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1C18 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects RUNX1/AML1. It targets an epitope within 22 amino acids from the internal region.

Immunogen

KLH-conjugated linear peptide corresponding to 22 amino acids from the internal region of human Runt-related transcription factor 1 (RUNX1).

Application

Quality Control Testing

Evaluated by Western Blotting in RAW264.7 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected RUNX1 in RAW264.7 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected RUNX1 in Rat Thymus lysates.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected RUNX1 in Jurkat cells.

Affinity Binding Assay: A representative lot of this antibody bound RUNX1 peptide with a KD of 2.5 x 10-7 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Runt-related transcription factor 1 (UniProt: Q01196; also known as Acute myeloid leukemia 1 protein, Core-binding factor subunit alpha-2, CBF-alpha-2, Oncogene AML-1, Polyomavirus enhancer-binding protein 2 alpha B subunit, PEA2-alpha B, PEBP2-alpha B, SL3-3 enhancer factor 1 alpha B subunit, SL3/AKV core-binding factor alpha B subunit) is encoded by the RUNX1 (also known as AML1, CBFA2) gene (Gene ID: 861) in human. RUNX members are shown to modulate the transcription of their target genes through recognizing the core consensus binding sequence 5′-TGTGGT-3′. RUNX1 is a core-binding transcription factor that is reported to be essential for the establishment of definitive hematopoiesis in vertebrates. It is expressed in all tissues except brain and heart. Its highest levels are observed in thymus, bone marrow, and peripheral blood. RUNX1 forms the heterodimeric complex core-binding factor (CBF) with CBFB and controls the expression of its target genes involved in hematopoietic differentiation, ribosome biogenesis, cell cycle regulation, and p53 and transforming growth factor β signaling pathways. RUNX1 binds DNA as a monomer through its Runt domain (aa 50-178) and its DNA-binding is increased by heterodimerization. RUNX1 is phosphorylated on serine 249 and 276 and threonine 273 by homeodomain interacting protein kinase 2 (HIPK2) when associated with CBFB and DNA. This phosphorylation promotes subsequent EP300 phosphorylation. RUNX1 gene is one of the most frequently mutated genes in a variety of hematological malignancies. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Sood, R., et al. (2017). Blood. 129(15); 2070-2082 Wee, H-J., et al. (2008). Blood. 112(9); 3777-3787).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service