Skip to Content
MilliporeSigma

MAB5460

Anti-Nkx2.1 Antibody, clone 8G7-G3-1

ascites fluid, clone 8G7-G3-1, Chemicon®

Synonym(s):

TTF-1, Thyroid Transcription Factor-1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
8G7-G3-1, monoclonal
Application:
IHC, WB
Citations:
17
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

8G7-G3-1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NKX2-1(7080)

Immunogen

Recombinant rat Nkx2.1.

Application

This Anti-Nkx2.1 Antibody is validated for use in WB, IH(P) for the detection of Nkx2.1.
Western blot. The antibody reacts with a 40 kDa band in immunoblots of nuclear extracts or whole cell lysates from the Nkx2.1 positive cell lines MLE15, H441-4, H345 and rat type II pmeumocyte cells.

Immunohistochemistry. The antibody can be used on formalin-fixed paraffin embedded tissue. Heat induced epitope retrieval is recommended.

Optimal working dilutions must be determined by end user.

Biochem/physiol Actions

Nkx2.1 (Thyroid Transcription Factor-1, TTF-1).

Analysis Note

Tested

Other Notes

Please refer to lot specific datasheet.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library