Skip to Content
MilliporeSigma
All Photos(10)

Key Documents

ZRB1925

Sigma-Aldrich

Anti-Neuropilin-1 Antibody, clone 1H21 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1H21, recombinant monoclonal

description

1H21 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 103.13 kDa
observed mol wt ~130 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

canine, feline, monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... NRP1(8829)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1H21 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Neuropilin-1. It targets an epitope within 19 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 19 amino acids from the C-terminal region of human Neuropilin-1.

Application

Quality Control Testing

Evaluated by Western Blotting in Human umbilical vascular endothelial cells (HUVEC) lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected Neuropilin-1 in Human umbilical vascular endothelial cells (HUVEC) lysate.

Tested applications

Flow Cytometry Analysis: 0.1 μg from a representative lot detected Neuropilin-1 in one million Human umbilical vascular endothelial cells (HUVEC).

Affinity Binding Assay: A representative lot of this antibody bound Neuropilin-1 peptide with a KD of 2.2 x 10-8 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Neuropilin-1 in Human umbilical vascular endothelial cells (HUVEC).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Neuropilin-1 (UniProt: O14786; also known as Vascular endothelial cell growth factor 165 receptor, CD304) is encoded by the NRP1 (also known as NRP, VEGF165R) gene (Gene ID: 8829) in human. Neuropilins are highly conserved, non-tyrosine kinase, single-transmembrane glycoproteins that serve as co-receptors for various molecules, including semaphorins and vascular endothelial growth factors (VEGF). Neuropilin-1 is a single-pass type I, homodimeric, membrane glycoprotein that is synthesized with a signal peptide (aa 1-21), which is subsequently cleaved off to generate the mature form that contains a long extracellular domain (aa 22-856), a transmembrane domain (aa 857-879), and a cytoplasmic domain (aa 880-923). It acts as a receptor involved in the development of the cardiovascular system, angiogenesis, and formation of certain neuronal circuits. It is also reported to mediate the chemorepulsant activity of semaphorins. Neuropilin-1 has four well conserved domains: CUB (a1/a2; aa 27-141 and 147-265) domain, F5/8 (b1/b2; aa 275-424 and 431-583) domain, MAM domain (aa 645-811), and another domain that contains a transmembrane and a short cytoplasmic region. CUB domains mediate binding to semaphorins, while the tandem F5/8 domains are responsible for VEGF and heparin binding and recognizes a C-end role (CendR) motif R/KXXR/K on its ligands. The presence of the CUB (a1a2) domain is reported to enhance VEGF165 binding to F5/8 (b1b2) domain. The MAM domain is reported to mediate semaphorin signal transduction to Plexin-A. Neuropilin-1 has been recognized as an additional cellular mediator that facilitates the entry of SARS-CoV-2 into host cells. It binds to furin-cleaved spike protein and significantly potentiates SARS-CoV-2 infectivity. This effect can be blocked by monoclonal antibodies against the extracellular F5/8 (b1/b2) domain of Neuropilin-1 or with selective Neuropilin antagonists that bind to CendR binding b1 pocket. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Kyrou, I., et al. (2021). Signal Trans. Targeted Ther. 6; Article number: 21; Daly, JL., et al. (2020). Science. 370(6518); 861-865; Mamluk, R., et al. (2002). J. Biol. Chem. 277(27); 24818-24825; Soker, S., et al. (1998). Cell. 92(6); 735-745).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service