Skip to Content
MilliporeSigma
All Photos(14)

Documents

ZRB1233

Sigma-Aldrich

Anti-Caspase-1 (p20) Antibody, clone 2G4 , ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

CASP-1, EC:3.4.22.36, ICE, IL-1 beta-converting enzyme, IL-1BC, Interleukin-1 beta convertase, Interleukin-1 beta-converting enzyme, p45

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2G4, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

observed mol wt ~20 kDa
observed mol wt ~45 kDa

species reactivity

human, mouse, rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CASP1(834)
mouse ... Casp1(12362)
rat ... Casp1(25166)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 2G4 is a ZooMAb rabbit recombinant monoclonal antibody that detects Caspase-1 proenzyme and the p20 subunit. It targets an epitope within 21 amino acids from the C-terminal region of p20 subunit.

Immunogen

KLH-conjugated linear peptide corresponding to 21 amino acids from the p20 subunit of human Caspase-1.

Application

Anti-Caspase-1 (p20), clone 2G4 ZooMAb, Cat. No. ZRB1233, is a recombinant Rabbit monoclonal antibody that targets Caspase-1 proenzyme and p20 subunit and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Caspase-1 proenzyme and p20 subunit in rat spleen tissue lysate.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Caspase-1 (p20) in NIH 3T3 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Caspase-1 (p20) in human small intestine tissue sections.

Note: Actual optimal working dilutions must be determined by the end user as specimens and experimental conditions may vary.

Target description

Caspase-1 (UniProt: P29466; also known as EC:3.4.22.36, CASP-1, Interleukin-1 beta convertase, IL-1BC, Interleukin-1 beta-converting enzyme, ICE, IL-1 beta-converting enzyme, p45) is encoded by the CASP1 (also known as IL1BC, IL1BCE) gene (Gene ID: 834) inn human. Caspse-1 is a thiol protease that is synthesized as a single chain zymogen, which is further processed to create active caspase-1. The zymogen has two propeptides (aa 1-119 and 298-316), that are subsequently cleaved off to generate p20 (aa 120-297) and p10 (aa 317-404) subunits. The mature enzyme consists of two anti-parallel arranged heterodimers, each formed by a p20 and a p10 subunit. The p20 subunit can also form a heterodimer with the epsilon isoform which then has an inhibitory effect. It displays a strict requirement for an aspartic acid residue at position P1 and has a preferred cleavage sequence of Tyr-Val-Ala-Asp. Caspase-1 cleaves IL-1 beta between an aspartic acid and alanine, releasing the mature cytokine which is involved in a variety of inflammatory processes. It is an important component of the inflammasome that during DNA virus infection controls antiviral immunity through cleavage of cGMP-AMP synthase (cGAS), rendering it inactive. Caspase-1 is expressed in larger amounts in spleen and lung, but is also detected in most other tissues, except for the brain. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Wang, Y., et al. (2017). Immunity. 46(3); 393-404).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

30 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mei-Hsin Hsu et al.
Pharmaceuticals (Basel, Switzerland), 16(6) (2023-06-28)
Endothelial dysfunction is characterized by disturbances in nitric oxide (NO) bioavailability and increased circulating asymmetric dimethylarginine (ADMA) due to the enormous release of free radicals. Increased circulating ADMA may cause endothelial dysfunction and a variety of clinical disorders, such as

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service