Skip to Content
MilliporeSigma
All Photos(16)

Documents

ZRB1837

Sigma-Aldrich

Anti-Phospho-c-Jun (Ser73) Antibody, clone 2B14 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Transcription factor AP-1 (UniProt: P05412; also known as Activator protein 1, AP1, Proto-oncogene c-Jun, V-jun avian sarcoma virus 17 oncogene homolog, p39

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2B14, recombinant monoclonal

description

2B14 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 35.68 kDa
observed mol wt ~40 kDa

purified by

using Protein A

species reactivity

mouse, human

species reactivity (predicted by homology)

monkey, canine, bovine, rat, horse, zebrafish, porcine

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunohistochemistry: suitable
inhibition assay: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

phosphorylation (pSer73)

Gene Information

human ... JUN(3725)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2B14 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects c-Jun phosphorylated on serine 73.

Immunogen

KLH-conjugated linear peptide corresponding to 13 amino acids surrounding phosphoserine 73 of human c-Jun.

Application

Quality Control Testing

Evaluated by Western Blotting in lysates from NIH3T3 cells treated with UV light.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected Phospho-c-Jun (Ser73) in lysates from NIH3T3 cells treated with UV light (50 mJoules followed by 5 h recovery), but not in untreated cells.

Tested Applications

Flow Cytometry Analysis: 0.1 μg from a representative lot detected Phospho-c-Jun (Ser73) in one million HeLa cells treated with UV light

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Phospho-c-Jun (Ser73) in human colon tissue sections.

Peptide Inhibition Assay: Target band detection in lysates from NIH3T3 cells was prevented by preblocking of a representative lot with the Phospho-c-Jun (Ser73) peptide, but not the corresponding non-phosphopeptide.

Affinity Binding Assay:: A representative lot of this antibody bound Phospho-c-Jun (Ser73) peptide with a KD of 4.5 x 10-7 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected Phospho-c-Jun (Ser73) in HeLa cells treated with UV light.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Transcription factor AP-1 (UniProt: P05412; also known as Activator protein 1, AP1, Proto-oncogene c-Jun, V-jun avian sarcoma virus 17 oncogene homolog, p39) is encoded by the JUN gene (Gene ID: 3725) in human. c-Jun is a transcription factor that forms homodimers with JunB and JunD or heterodimers with Fos family members. These homodimers and heterodimers bind to specific DNA sequences, known as 12-Otetradecanoylphorbol-13-acetate response elements (TRE), in the promoter regions of target genes and activate transcription. c-Jun recognizes and binds to the enhancer heptamer motif 5′-TGA[CG]TCA-3′. It is involved in numerous cellular activities, including cell proliferation, apoptosis, survival, tumorigenesis, and tissue morphogenesis. c-Jun is phosphorylated by CaMK4 and DNA dependent protein kinase, which enhances its transcriptional activity. When phosphorylated by HIPK3, it promotes the activity of Nuclear Receptor Subfamily 5 Group A Member 1 (NR5A) that leads to increased steroidogenic gene expression upon stimulation of cAMP signaling pathway. It also undergoes phosphorylation at serine 243 by DYRK2 that primes it for phosphorylation by GSK-3β at threonine 239, serine 243, and serine 249. These phosphorylations reduce its DNA binding ability and allows its ubiquitination and degradation. On the other hand, when ERK activates p70S6 kinase, the activated p70S6 kinase phosphorylates GSK3β at serine 21 that leads to dephosphorylation and increased DNA binding activity of c-Jun. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Lukey, MJ., et al. (2016). Nat. Commun. 7; Article 11321; Qinghang Meng, Q., and Xia, Y. (2011). Protein Cell. 2(11); 889-898; Zhang, Y., et al. (2007). BMC Cancer. 7; Article 145; Lan, HC., et al. (2007). Mol. Cell Biol. 27(6); 2027-2036; Nateri, AS., et al. (2004). Science. 303(5662); 1374-1378).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service