Skip to Content
MilliporeSigma
All Photos(12)

Key Documents

ZRB1721

Sigma-Aldrich

Anti-EGR1 Antibody, clone 1F22 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Early growth response protein 1;AT225;Nerve growth factor-induced protein A;NGFI-A;Transcription factor ETR103;Transcription factor Zif268;Zinc finger protein 225;Zinc finger protein Krox-24

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1F22, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 57.51 kDa
observed mol wt ~80 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... EGR1(1958)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1F22 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Early growth response protein 1 (EGR1). It targets an epitope within 23 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 23 amino acids from the N-terminal half of human Early growth response protein 1 (EGR1).

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected EGR1 in HeLa cell lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected EGR1 in PC3 cell lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected EGR1 in human prostate tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected EGR1 in PC3 cells.

Affinity Binding Assay: A representative lot of this antibody bound EGR1 with a KD of 1.3 x 10-8 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Early growth response protein 1 (UniProt: P18146; also known as EGR-1, AT225, Nerve growth factor-induced protein A, NGFI-A, Transcription factor ETR103, Transcription factor Zif268, Zinc finger protein 225, Zinc finger protein Krox-24) is encoded by the EGR1 (also known as KROX24, ZNF225) gene (Gene ID: 1958) in human. EGR1 is a member of the EGR C2H2-type zinc-finger protein family that serves as a transcriptional regulator. It recognizes and binds to the DNA sequence 5′-GCG(T/G)GGGCG-3′(EGR-site) in the promoter region of target genes and regulates their transcription. It contains three C2H2-type Zinc fingers (a 338-362, 368-390, 396-418). The first, most N-terminal zinc finger binds to the 3′-GCG motif, the middle zinc finger interacts with the central TGG motif, and the C-terminal zinc finger binds to the 5′-GCG motif. It is shown to binds double-stranded target DNA, irrespective of the cytosine methylation status. It displays reduced affinity for target DNA where cytosines have been oxidized to 5-hydroxymethylcytosine and does not bind target DNA where cytosines have been oxidized to 5-formylcytosine or 5-carboxylcytosine. EGR1 plays an important role in regulating the response to growth factors, DNA damage, and ischemia. It is reported to activate the expression of p53/TP53 and TGFB1, and thereby prevents tumor formation and is shown to be essential for normal progress through mitosis and normal proliferation of hepatocytes after partial hepatectomy. It is also regulates the expression of proteins such as IL-1b and CXCL2 that are involved in inflammatory processes and development of tissue damage after ischemia. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Zandarashvili, L., et al. (2015). FEBS Lett. 589(15); 1748-1753; Hashimoto, H., et al. (2014). Genes Dev. 28(20); 2304-2313).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

30 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service