Skip to Content
MilliporeSigma
All Photos(12)

Key Documents

ZRB1073

Sigma-Aldrich

Anti-Stathmin-2 Antibody, clone 1B3 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Protein SCG10, Superior cervical ganglion-10 protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

clone

1B3, recombinant monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 20.83 kDa
observed mol wt ~21 kDa

species reactivity

human, rat, mouse

species reactivity (predicted by homology)

monkey, bovine

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1B3 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Stathmin-2. It targets an epitope within 20 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the N-terminal region of human Stathmin-2.

Application

Quality Control Testing
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Stathmin-2 in Neuro-2A lysate.Tested applicationsWestern Blotting Analysis: A 1:1,000 dilution from a representative lot detected Stathmin-2 in PC12 and SH-SY5Y lysates.Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Stathmin-2 in SH-SY5Y cells. Affinity Binding Assay: A representative lot of this antibody bound Stathmin-2 peptide with a KD of 1.5 x 10-8 in an affinity binding assay. Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Stathmin-2 in human cerebral cortex tissue sections.Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Stathmin-2, clone 1B3 ZooMAb, Cat. No. ZRB1073, is a recombinant Rabbit monoclonal antibody that specifically targets Stathmin-2 and is tested for use in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry, and Western Blotting.

Target description

Stathmin-2 (UniProt: Q93045; also known as Superior cervical ganglion-10 protein, Protein SCG10) is encoded by the STMN2 (also known as SCG10, SCGN10) gene (Gene ID: 11075) in human. Stathmin-2 is a neurophosphoprotein that acts as a regulator of microtubule stability. It is a peripheral membrane protein that is also found in Golgi and endosomes. Its membrane attachment domain is localized in amino acids 1-26 and phosphorylation/regulatory domain is in amino acids 39-96. When phosphorylated by MAPK8, it stabilizes microtubules and consequently controls neurite length in cortical neurons. Its N-terminal palmitoylation promotes specific anchoring to the cytosolic leaflet of Golgi membranes and subsequent vesicular trafficking along dendrites and axons. In the developing brain, it is reported to negatively regulate the rate of exit from multipolar stage and retards radial migration from the ventricular zone. Stathmin-2 is considered as novel regulator of glucagon secretion and its expression has been reported in murine pancreatic alpha cells where its overexpression increases localization of glucagon into the endosomal-lysosomal compartment and depletion results in reduced endosomal localization of glucagon. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Asadi, F., and Dhanvantari, S. (2020). Front Endocrinol.11; 29).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

30 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service