Skip to Content
MilliporeSigma
All Photos(13)

Documents

ZRB1521

Sigma-Aldrich

Anti-MDR-1 Antibody, clone 1O12 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

ATP-dependent translocase ABCB1, CD243, Multidrug resistance protein 1, P-glycoprotein 1, Phospholipid transporter ABCB1, also known as ATP-binding cassette sub-family B member 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1O12, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 141.48 kDa
observed mol wt ~150 kDa

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... ABCB1(5243)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1O12 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects MDR1 protein. It targets an epitope within 22 amino acids from internal, cytoplasmic loop region.

Immunogen

KLH-conjugated linear peptide corresponding to 22 amino acids from the internal cytoplasmic loop region of human Multidrug resistance protein 1 (MDR-1).

Application

Anti-MDR-1, clone 1O12 ZooMAb, Cat. No. ZRB1521, is a recombinant Rabbit monoclonal antibody that targets MDR-1 and is tested in Affinity Binding Assay, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected MDR-1 in HepG2 and NIH 3T3 cells.

Affinity Binding Assay: A representative lot bound MDR-1 peptide with a KD of 3.5 x 10-7 in an affinity binding assay.

Flow Cytometry Analysis: 0.01 μg from a representative lot detected MDR-1 in HepG2 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected MDR-1 in human adrenal gland tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Multidrug resistance protein 1 (UniProt: P08183; also known as ATP-binding cassette sub-family B member 1, ATP-dependent translocase ABCB1, P-glycoprotein 1, Phospholipid transporter ABCB1, CD243; MDR1) is encoded by the ABCB1 (also known as MDR1, PGY1) gene (Gene ID: 5243) in human. In tumor cells, the classical MDR phenotype is conferred by MDR1 that is expressed in about 50% of human cancers. MDR1 is a multi-pass membrane N-linked glycoprotein that translocates drugs and phospholipids across the membrane. It contains 6 extracellular domains, 12 transmembrane domains, and 7 cytoplasmic domains. Its ABC transporter domains are localized to amino acids 392-628 and 1035-1273.MDR1 expression has been reported in liver, kidney, small intestine, and brain. It catalyzes the flop of phospholipids from the cytoplasmic to the exoplasmic leaflet of the apical membrane. It participates mainly to the flop of phosphatidylcholine, phosphatidyl-ethanolamine, beta-D-glucosylceramides and sphingomyelins. It serves as an energy-dependent efflux pump that is responsible for decreased drug accumulation in multidrug-resistant cells. Overexpression of MDR1 is shown to be of prognostic value in leukemia, neuroblastoma and sarcoma and its inhibition can restore chemosensitivity in tumor cells. Mutations in ABCB1 gene has been linked to inflammatory bowel disease 13, a chronic, relapsing inflammation of the gastrointestinal tract with a complex etiology. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Perez, J., et al. (2011). Anticancer Res. 31(9); 2813-2820; Kaszubiak, A., et al. (2007). Int. J. Oncology 31(2); 419-430).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

0.3 mg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service