Skip to Content
MilliporeSigma
All Photos(14)

Documents

ZRB1304

Sigma-Aldrich

Anti-CRBN Antibody, clone 3I23 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Anti-MRT2, Anti-MRT2A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3I23, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 50.55 kDa
observed mol wt ~50 kDa

species reactivity

rat, mouse, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminus

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 3I23 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Cereblon. It targets an epitope within 19 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 19 amino acids from the N-terminal region of human Cereblon.

Application

Quality Control Testing

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected CRBN in Jurkat cell lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Cereblon (CRBN) in NIH3T3, PC3, and C6 cell lysates.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected Cereblon (CRBN) in human cerebral cortex tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Cereblon (CRBN) in PC3 and Jurkat cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-CRBN, clone 3I23 ZooMAb®, Cat. No. ZRB1304, is a recombinant Rabbi+J3:J17t monoclonal antibody that specifically targets Cereblon (CRBN) and is tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.

Target description

Protein cereblon (UniProt: Q96SW2) is encoded by the CRBN (also known s AD-006) gene (Gene ID: 51185) in human. Cereblon is peripheral membrane protein that is widely expressed with highest levels found in the brain. It is reported to play a protective role against DNA damage-induced apoptosis in cell lines and primary cells. It interacts with DNA-binding protein 1 (DDB1), Cullin 4 (Cul4A or Cul4B), and regulator of Cullins 1 (RoC1) to form the functional E3 ubiquitin ligase complex where cereblon functions as a substrate receptor of E3 ubiquitin ligase complex, which mediates the ubiquitination and subsequent proteasomal degradation of target proteins, such as MEIS2. The cullin-4 protein provides a scaffold for assembling RING box-domain protein (RBX1) and the adaptor DDB1 to form a CRL4 E3 complex, where RBX1 serves as the docking site for the activated E2 protein. The DDB1 protein recruits substrate-specificity receptors or DDB1-cullin 4-associated factors (DCAFs) to form the substrate-presenting side of the Cullin-RING E3 ubiquitin ligases complex. Cereblon has been recognized as the primary teratogenic target of thalidomide and its analogs. These drugs exert their antiproliferative effects on multiple myeloma cells and immunomodulatory effects on T cells by directly binding cereblon and promoting recruitment of the hematopoietic zinc-finger transcription factors Ikaros (IKZF1) and Aiolos (IKZF3), leading to their ubiquitination and proteasomal degradation. Mutations in CRBN gene have been linked to mental retardation. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Shi, Q., and Chen, L. (2017). J. Immunol. Res. 2017; Article 9130608; Zhou, L., and Xu, G. (2019). Cell Death Dis. 10; Article 69).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service