Skip to Content
MilliporeSigma
All Photos(9)

Key Documents

ZRB1300

Sigma-Aldrich

Anti-SCN2A Antibody, clone 3F19 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3F19, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 227.98 kDa
observed mol wt ~260 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 3F19 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Sodium channel protein type 2 subunit alpha (SCN2A). It targets an epitope within 23 amino acids from the C-terminal, cytoplasmic domain.

Immunogen

KLH-conjugated linear peptide corresponding to 23 amino acids from the C-terminal, cytoplasmic domain of human Sodium channel protein type 2 subunit alpha (SCN2A).

Application

Quality Control Testing

Evaluated by Western Blotting in Human brain tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected SCN2A in Human brain tissue lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected SCN2A in Human hippocampus membrane preparations.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected SCN2A in K562 cells (not permeabilized).

Affinity Binding Assay: A representative lot of this antibody bound SCN2A in SCN2A Sodium Channel with a KD of 1.2 x 10-8 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-SCN2A, clone 3F19 ZooMAb®, Cat. No. ZRB1300, is a recombinant Rabbit monoclonal antibody that detects SCN2A and is tested for use in Affinity Binding Assay, Immunocytochemistry, and Western Blotting.

Target description

Sodium channel protein type 2 subunit alpha (UniProt: Q99250; also known as HBSC II, Sodium channel protein brain II subunit alpha, Sodium channel protein type II subunit alpha, Voltage-gated sodium channel subunit alpha Nav1.2, SCN2A) is encoded by the SCN2A (also known as NAC2, SCN2A1, SCN2A2) gene (Gene ID: 6326) in human. Sodium channels are integral plasma membrane proteins that conduct Na+ transport across the plasma membrane. They are classified as either voltage-gated or ligand-gated based on the nature of the triggering mechanism for channel activation. SCN2A is a multi-pass membrane glycoprotein with 13 cytoplasmic domains, 16 extracellular domains, and 24 transmembrane domains, and It mediates the voltage-dependent sodium ion permeability of excitable membranes. It forms a sodium-selective channel through which Na+ ions may pass in accordance with their electrochemical gradient. Mutations in SCN2A gene are known to cause infantile epileptic encephalopathy that is characterized by infantile-onset seizures, before 12 months of age, followed by neurodevelopmental delay. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Sanders, SJ., et al. (2018). Trends Neurosci. 41(7); 442-456).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb® formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service