Key Documents
Z215899
Solid PTFE stopper
joint: ST/NS 10/30
Synonym(s):
flask stopper, lab stopper, stoppers
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
Joint
joint: ST/NS10/30
manufacturer/tradename
Cowie 010.110
General description
Machined from solid PTFE with trapezoidal seals for medium to high vacuum performance up to 280°C (540°F).