Skip to Content
MilliporeSigma

Skip To

PRS3463

Anti-Caspase-1 (ab2) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Caspase 1, apoptosis-related cysteine peptidase (Interleukin 1, beta, convertase), Anti-ICE, Anti-IL-1β converting enzyme

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Technique(s):
western blot: suitable, immunofluorescence: suitable, immunohistochemistry: suitable, indirect ELISA: suitable
Conjugate:
unconjugated
Application:
ELISA (i), IF, IHC, WB
Citations:
3
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: suitable, immunofluorescence: suitable, immunohistochemistry: suitable, indirect ELISA: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CASP1(834)

General description

Caspase-1 is also known as interleukin-1βconverting enzyme (ICE). It is located on chromosome 11q22.2. It has 10 exons, an N terminal CARD (caspase activation and recruitment domain), a large P20 subunit and a small P10 subunit. It is present in leukocytes, monocytes and epithelial cells.

Immunogen

a 15 amino acid peptide from near the middle of human Caspase-1.

Biochem/physiol Actions

Caspase-1 can transform the pro-inflammatory cytokines, interleukin-1β (IL-1β) and IL-18 into their active forms. It participates in pyroptosis. It is overexpressed in the aorta of coronary atherosclerosis patients.It helps in host cell survival by inducing membrane biogenesis to restore the impairment caused by pore-forming toxins.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Other Notes

The action of this antibody can be blocked using blocking peptide SBP3463.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
WH0000834M1SAB5700660SAB4503271
description

affinity isolated antibody, buffered aqueous solution

description

clone 3D2, purified immunoglobulin, buffered aqueous solution

description

-

description

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

biological source

rabbit

biological source

mouse

biological source

rabbit

biological source

rabbit

shipped in

dry ice

shipped in

dry ice

shipped in

wet ice

shipped in

wet ice

technique(s)

immunofluorescence: suitable, indirect ELISA: suitable, western blot: suitable, immunohistochemistry: suitable

technique(s)

indirect ELISA: suitable, western blot: 1-5 μg/mL, immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

technique(s)

immunofluorescence: 1:50-1:200, western blot: 1:500-1:2000, immunohistochemistry: 1:50-1:200, immunoprecipitation (IP): 1:50-1:100

technique(s)

immunohistochemistry: 1:50-1:100, ELISA: 1:20000, western blot: 1:500-1:1000


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters