Skip to Content
MilliporeSigma
All Photos(12)

Key Documents

ZRB1131

Sigma-Aldrich

Anti-LAMP-1 Antibody, clone 2E9 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2E9, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 44.88 kDa
observed mol wt ~110 kDa

species reactivity

monkey, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 2E9 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects LAMP-1. It targets an epitope within 14 amino acids from the N-terminal, lumenal domain.

Immunogen

KLH-conjugated linear peptide corresponding to 14 amino acids from the N-terminal, lumenal domain of human LAMP-1.

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected LAMP-1 in HeLa cell lysate.

Tested applications


Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected LAMP-1 in K562, Jurkat, and Cos-7 cell lysates.

Immunofluorescence Analysis: A 1:100 dilution from a representative lot detected LAMP-1 in human placenta tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected LAMP-1 in human placenta tissue sections.

Flow Cytometry Analysis: 0.1 μg from a representative lot detected LAMP-1 in one million K562 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-LAMP-1, clone 2E9 ZooMAb®, Cat. No. ZRB1131, is a recombinant Rabbit monoclonal antibody that specifically detects LAMP-1 and is tested for use in Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), and Western Blotting.

Target description

Lysosome-associated membrane glycoprotein 1 (UniProt: P11279; also known as LAMP-1, Lysosome-associated membrane protein 1, CD107 antigen-like family member A, CD107a) is encoded by the LAMP1 gene (Gene ID: 3916) in human. LAMP-1 and LAMP-2 are two highly glycosylated proteins found in the lysosomal membrane. They may contribute up to 50% of all proteins present in the lysosomal membranes. They are known to maintain the lysosomal acidification and lysosomal membrane integrity. Extensive N-glycosylation on LAMP-1 and 2 to protects them from the lysosomal proteolysis. However, higher levels of poly-N-acetyllactosamines on LAMP-1 and 2 have been correlated with decreased cellular differentiation and enhanced metastatic potential. Over-expression of LAMP-1 has also been implicated in breast cancer development and progression. LAMP-1 is a single-pass type I transmembrane protein that is synthesized with a signal peptide (aa 1-28), which is subsequently cleaved off to generate the mature form that contains a large lumenal domain (aa 29-382), a transmembrane domain (aa 383-405), and a short C-terminal cytoplasmic tail (aa 406-417). LAMP-1 presents carbohydrate ligands to selectins and is also reported to act as a receptor for Lassa virus protein. It is reported to shuttle between lysosomes, endosomes, and the plasma membrane. Two isoforms of LAMP-1 have been described that are produced by alternative splicing. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Wang, Q., et al. (2017). Oncol. Lett. 14(4); 4729 - 4735; Tan, K-P., et al. (2016). Cell Death Dis. 7, e2347).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

30 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service